Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) cannot be used in connection with option K 1 MAX. LOADING 4 KG 1 * 1 items
Catalog # MEMMB29726UF260TS
Supplier: MEMMERT