Order Entry
Denmark
ContactUsLinkComponent
MISA Standard 6 - Transition Metals - 13 components: 100mg/l each of Cd Co Cu Cr Fe Pb Mn Hg Ni Ag Tl V Zn in HNO3 10% 1 * 100 mL
Catalog # ACSDE579.K1.10N.L1
Supplier:  CPACHEM
New Product
MISA Standard 6 - Transition Metals - 13 components: 100mg/l each of Cd Co Cu Cr Fe Pb Mn Hg Ni Ag Tl V Zn in HNO3 10% 1 * 100 mL
Catalog # ACSDE579.K1.10N.L1
Supplier:  CPACHEM