Order Entry
Denmark
ContactUsLinkComponent
Mineral Oil Heavy CAS:8042-47-5 EC:232-455-8 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 mL
Catalog # ACSDSB65049.1000
Supplier:  CPACHEM
New Product
Mineral Oil Heavy CAS:8042-47-5 EC:232-455-8 CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 mL
Catalog # ACSDSB65049.1000
Supplier:  CPACHEM