Muffle furnace l9/11/skm/c550 tmax1100 flap door and thermocouple hole in the furnace door 1 * 1 items
NABELM095K1ANOY
New Product
Muffle furnace l9/11/skm/c550 tmax1100 flap door and thermocouple hole in the furnace door 1 * 1 items
NABELM095K1ANOY
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection