Canister S60/61. 5l. made of static dissipative PE-HD-EX. with thread S60/61 (DIN60/61). straight neck. dimensions: L 195 x W 165 x H 230 mm 1 * 1 items
Catalog # BOHLR231-05
Supplier: Bohlender
New Product
Canister S60/61. 5l. made of static dissipative PE-HD-EX. with thread S60/61 (DIN60/61). straight neck. dimensions: L 195 x W 165 x H 230 mm 1 * 1 items
Catalog # BOHLR231-05
Supplier: Bohlender
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Jerricans, narrow neck, HDPE, transparent or blue
Supplier: BURKLE
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection