11 components: 100mg/l each of Li Mo Ni Sb Be Co Pb V Sn Mg Ti in HNO3 5% 1 * 100 mL
Catalog # CMPLRM113432L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
11 components: 100mg/l each of Li Mo Ni Sb Be Co Pb V Sn Mg Ti in HNO3 5% 1 * 100 mL
Catalog # CMPLRM113432L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
4 components: 100mg/l each of Ag As Cd Hg in HNO3 5% 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB