Order Entry
Denmark
ContactUsLinkComponent
Long description: High temperature furnace LHT 08/17 with controller P580, protective gas connection and over-temperature limiter, chamber volume: 8 L, max. Temperature: 1750 °C 1 * 1 items
  NABELHT827WN1GT
New Product
Long description: High temperature furnace LHT 08/17 with controller P580, protective gas connection and over-temperature limiter, chamber volume: 8 L, max. Temperature: 1750 °C 1 * 1 items
  NABELHT827WN1GT