Antibiotic Sensitivity Discs
Ketoconazole, 10 mcg, 50
HIMESD224-5X50DS
: HIMEDIA
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
- Pk:50
- Description:Ketoconazole, 10 mcg