Antibiotic Sensitivity Discs
Doxycycline hydrochloride, 30 mcg, 50
Catalog # HIMESD012-5X50DS
Supplier: HIMEDIA
Jump to:
Product Details & Documents
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
Specifications
Specifications
- Pk:50
- Description:Doxycycline hydrochloride, 30 mcg