Order Entry
Denmark
ContactUsLinkComponent
Glass shaft, sensor silver cap 4 mm ?, sulfidized, plug head, length 120 mm, 12 mm ?, -5...+100 °C Replaces the Ag 1100 + sulfidizing 285901057 1 * 1 items
Katalog # SCOI285103620
Supplier:  SI Analytics
New Product
Glass shaft, sensor silver cap 4 mm ?, sulfidized, plug head, length 120 mm, 12 mm ?, -5...+100 °C Replaces the Ag 1100 + sulfidizing 285901057 1 * 1 items
Katalog # SCOI285103620
Supplier:  SI Analytics