Order Entry
Denmark
Orders LinkContactUsLinkComponent
Selective Supplements

Selective Supplements

Blaser-Wang selective supplement for the selective isolation of Campylobacter species, 1
Catalog # HIMEFD006-5X5VL
Supplier:   HIMEDIA
Selective Supplements
Catalog # HIMEFD006-5X5VL
Supplier:  HIMEDIA
CAS Number:  
Selective Supplements

Blaser-Wang selective supplement for the selective isolation of Campylobacter species, 1

CAS Number:  
Price
1.990,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Antibiotic supplement recommended for the selective isolation of various microorganisms.

  • Store at 2...8 °C
Specifications

Specifications

  • Pk:
    1
  • Description:
    Blaser-Wang selective supplement for the selective isolation of <i>Campylobacter</i> <i>species</i>

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.