PE Medium Adhesion Cleanroom Tape
Product Details & Documents
Particle-free, residue-free tapes.
- Medium adhesion polyethylene tapes
- Good alternative to vinyl tapes
- UltraClean material
These general use and cleanroom construction tapes bond quickly to all surfaces. The adhesive strength is virtually identical to vinyl tapes. The polyethylene backing can be written on with a cleanroom pen, and it can be torn by hand.
UltraTape 1154/0154 is also recommended for in-process applications due to its low outgassing and metal properties. UltraTape 1112 has the same adhesive system (but different ratio of ingredients), and a medium density as opposed to a low density polyethylene backing.
Specifications
- Width:—
- Colour:Yellow
- Description:Ultratape 112, acrylic adhesive material
- Length:36 yards
- Seq:05