Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hand held tool and covers

Hand held tool and covers

PolyMesh™ slip cover, 100% polyester with mesh, foam interior, 152×102 mm, Polyester
Catalog # MICRMZSM68-6S
Supplier:   MICRONOVA
Hand held tool and covers
Catalog # MICRMZSM68-6S
Supplier:  MICRONOVA
Hand held tool and covers
Hand held tool and covers

PolyMesh™ slip cover, 100% polyester with mesh, foam interior, 152×102 mm, Polyester

Price
4.220,00 kr
Log in for availability
1 * 24 Items
Product Details & Documents

The hand held tool conforms to the hand and works with a choice of M-Zone hand held mop covers to ergonomically clean and scrub surfaces for a complete sanitization of small equipment and production surfaces.

  • Ergonomic cleaning for small equipment and surfaces
  • Lightweight, forms to hand
  • Available in polypropylene, recyclable plastic, or autoclavable polyketone adapters
  • Irradiated NovaPoly™ Cover is 100% smooth knit polyester, matches traditional cleanroom wipe fabrics, and provides excellent disinfectant dispersal
  • PolyMesh™ Cover is 100% polyester mesh, with foam interior, for scrubbing
  • Covers compatible with alcohol and strong disinfectants and are launderable for reuse
  • Autoclavable

The tool adapter is available in polypropylene, recyclable plastic, or an autoclavable polyketone. The slip-on covers are available in smooth polyester or polyester mesh fabric, to meet a variety of cleaning needs.

Specifications
  • Description:
    PolyMesh™ slip cover, 100% polyester with mesh, foam interior
  • L×W:
    152×102 mm
  • Material:
    Polyester
  • Seq:
    2