FAST FLEX KIT 2 XYLENE 1 * 3 items
Catalog # SHAN9802
Supplier: Epredia
New Product
Frequently Bought Together
Control slides, ISOSLIDE®
Supplier: Merck
Acid red 27 ≥85-95% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
7 COMPONENTS 100MG/L EACH 1 * 10 Ampoul
Supplier: CUSTOM MADE CHEMICALS LAB
Solvent black 5, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
ALL-TRANS-RETINOL 0.1% IN METHANOL 1 * 10 Ampoul
Supplier: CUSTOM MADE CHEMICALS LAB
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
Supplier: Thermo Fisher Scientific
Bromocresol green-methyl red solution mixed indicator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Fast blue BB salt ≥80% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
SILVER NITRATE STD SOLUTION 0.100N 1 * 1 L
Supplier: Hach