Accessories for Cryo Boxes
Driver, PP, natural, 3×3 compartments, 1×121×27 mm, 8 g
Catalog # GLWAZP33
Supplier: GLW
Jump to:
Product Details & Documents
Specifications
- Description:Driver, PP, natural, 3×3 compartments, 1×121×27 mm, 8 g
- Seq:13
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



