Order Entry
Denmark
Orders LinkContactUsLinkComponent
Colour Standards ASTM

Colour Standards ASTM

ASTM 1 colour standard, single solution, 500 mL
Catalog # PROOALK-ASTM10
Colour Standards ASTM
Catalog # PROOALK-ASTM10
CAS Number:  
Colour Standards ASTM

ASTM 1 colour standard, single solution, 500 mL

CAS Number:  
Price
3.030,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    ASTM 1 colour standard, single solution
  • Cat. No.:
    PROOALK-ASTM10
  • Storage temperature:
    RT
  • Method:
    Colour analysis

Specifications