Order Entry
Denmark
ContactUsLinkComponent
8 components: Ni 100mg/l Cu 100mg/l Co 100mg/l Pb 40mg/l As 40mg/l Zn 40mg/l Cd 20mg/l Hg 20mg/l in HNO3 5% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Catalog # ACSDRM107895L1
Supplier:  CPACHEM
New Product
8 components: Ni 100mg/l Cu 100mg/l Co 100mg/l Pb 40mg/l As 40mg/l Zn 40mg/l Cd 20mg/l Hg 20mg/l in HNO3 5% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Catalog # ACSDRM107895L1
Supplier:  CPACHEM