0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
[EN]REINFORCING BEAM FOR STANDARD 5/20/2 1 * 1 Items
HUPF0102102
:
HUPFER METALLWERKE
New Product
[EN]REINFORCING BEAM FOR STANDARD 5/20/2 1 * 1 Items
HUPF0102102
:
HUPFER METALLWERKE
New Product
Frequently Bought Together
VWR® Overalls, SMS
: VWR Collection
Laboratory trays
: Brand
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
VWR®, Nitril handsker, NITRILE
: VWR Collection
VWR®, Magnetstave, cylinderformede
: VWR Collection
Spatelsæt
: RSG
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Magnetstave, almindelige
: VWR Collection
VWR®, Engangsvejeskåle
: VWR Collection
VWR®, Engangsveje- og tørreskåle
: VWR Collection
Vejeskåle, engangs
: LUDI
Cross Brace, NORM5/25
: HUPFER METALLWERKE