14 COMPONENTS: 100MG/L EACH 1 * 100 mL
CMPLRM105224L1
New Product
Frequently Bought Together
Sodium hydroxide 30% (w/w) in aqueous solution, GPR RECTAPUR®
: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
NANOCOLOR® Photometric Tube Test Kits for Nitrate
: MACHEREY-NAGEL
Formalin 10%, Q Path®, buffered
: VWR Chemicals
VWR® Rota® Tri, pH indicator, 1 to 11
: VWR Chemicals