Order Entry
Denmark
Orders LinkContactUsLinkComponent
SoNa™ 520

SoNa™ 520

1 mg
Catalog # AATB21320
Supplier:   AAT Bioquest
SoNa™ 520
Catalog # AATB21320
Supplier:  AAT Bioquest
CAS Number:  
SoNa™ 520

1 mg

CAS Number:  
Price
5.220,00 kr
Log in for availability
1 * 1 mg
Product Details & Documents

SoNa™ 520 is a new sodium-sensitive fluorescent indicator dye used to detect sodium levels in cells and other biological samples.

  • Stable for 12 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:720.72 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications