Order Entry
Denmark
Orders LinkContactUsLinkComponent
SoNa™ 520

SoNa™ 520

10 x 50 µg, 50 µG
Catalog # AATB21321
Supplier:   AAT Bioquest
SoNa™ 520
Catalog # AATB21321
Supplier:  AAT Bioquest
CAS Number:  
SoNa™ 520

10 x 50 µg, 50 µG

CAS Number:  
Price
3.120,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

SoNa™ 520 is a new sodium-sensitive fluorescent indicator dye used to detect sodium levels in cells and other biological samples.

  • Stable for 12 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:720.72 g/mol
Specifications

Specifications

  • Pk:
    50 µG
  • Pk Info:
    10 x 50 µg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.