Order Entry
Denmark
ContactUsLinkComponent
PRIME-XV IS21 Supplement (50X) is a serum-free supplement optimized for use with PRIME-XV Neural Basal Medium to support maintenance and long- term culture of neuronal cells. This supplement is supplied as a 50X concentrated solution and should be di 1 * 10 mL
Catalog # SHBT91142
New Product
PRIME-XV IS21 Supplement (50X) is a serum-free supplement optimized for use with PRIME-XV Neural Basal Medium to support maintenance and long- term culture of neuronal cells. This supplement is supplied as a 50X concentrated solution and should be di 1 * 10 mL
Catalog # SHBT91142