Formaldehyde solution 4%
10 L
Catalog # AMPQ92555.1000-010
Supplier: Ampliqon A/S
Gå til:
Product Details & Documents
(10% Formaline solution)
Synonymer: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS nummer:50-00-0
- Formel:CH₂O
- MDL nummer:MFCD00003274
- Molecular Weight:30.03 g/mol
- Opbevaringstemperatur:Ambient
- EINECS:200-001-8
Specifikationer
Specifikationer
- Pk:10 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



