Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Formaldehyde solution 4%

1 L
Catalog # AMPQ92555.1000-001
Health
Irritant
Supplier:   Ampliqon A/S
Formaldehyde solution 4%
Catalog # AMPQ92555.1000-001
Supplier:  Ampliqon A/S
CAS Number:  50-00-0
Formaldehyde solution 4%
Formaldehyde solution 4%

1 L

CAS Number:  50-00-0
Price
3.400,00 kr
Log in for availability
1 * 1 L
Product Details & Documents

(10% Formaline solution)

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specifications

  • Pk:
    1 L

Specifications