O-RING F.SAFETY VALVE 1 * 1 items
Catalog # HIRSP9903204
Supplier: HIRSCHMANN
New Product
Frequently Bought Together
Mainboard 1 * 1 items
Supplier: MAPADA
GAS BALLAST TUBE 56,4 MM LONG 1 * 1 items
Supplier: VACUUBRAND
KIT FAN SYSTEM COMPLETE HTM Ø=180MM 230V 1 * 1 items
Supplier: Thermo Scientific
CLAMP BALLJOINT RV05.10 1 * 1 items
Supplier: IKA
Accessories for nitrogen analyser (VELP Scientifica)
Supplier: VELP SCIENTIFIC
Accessories for Laborota 20 control
Supplier: Heidolph Instruments GmbH & Co.KG
1000 and 1700 series Gastight® syringes for PSD/2 syringe pump
Supplier: HAMILTON BONADUZ
CLAMP RING 1 * 1 items
Supplier: VACUUBRAND
Kit Molecode A HALO 1 * 1 KIT
Supplier: ERLAB CAPTAIRE
CAROUSEL KIT, METAL FOR 10 BOTTLES Ø35MM 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach