NI - 1000 MG/L IN DILUTED HNO3 FOR ICP 1 * 30 mL
Catalog # CMPLC038.2NP.L03
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
NI - 1000 MG/L IN DILUTED HNO3 FOR ICP 1 * 30 mL
Catalog # CMPLC038.2NP.L03
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Chinese ink, green, Q Path® stain for histology
Supplier: VWR Chemicals
Spectroquant® PhotoCheck, Supelco®
Supplier: Merck
Test strips, nitrate , Reflectoquant®, Supelco®
Supplier: Merck
NANOCOLOR® Photometric Tube Test Kits for Nonionic/Anionic/Cationic Surfactants
Supplier: MACHEREY-NAGEL
Visual colour kits, pH indicator with colour card, Supelco®
Supplier: Merck
Powder pillows, sulphate, Sulfa 4
Supplier: Lovibond
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Supplier: Thermo Fisher Scientific
Sodium hydroxide ≥98.0% (by acidimetry), pellets ACS, ISO for analysis
Supplier: PanReac AppliChem