Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant PDGF-DD

Human Recombinant PDGF-DD

50 µG
Catalog # STMC78222.1
Marketsource
Human Recombinant PDGF-DD
Catalog # STMC78222.1
Marketsource
CAS Number:  
Human Recombinant PDGF-DD
Human Recombinant PDGF-DD

50 µG

CAS Number:  
Price
5.572,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

The platelet-derived growth factor (PDGF) family has five heparin-binding members that assemble into four homodimers (PDGF-AA, PDGF-BB, PDGF-CC, and PDGF-DD) and one heterodimer (PDGF-AB; Fretto et al.; Li and Eriksson). PDGF signals through the receptor tyrosine kinases PDGFRα and PDGFRβ. It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al.). PDGF is a potent mitogen for cells of mesenchymal origin, such as fibroblasts, glial cells, and vascular smooth muscle cells. PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.). PDGF-DD promotes growth and survival of renal artery smooth muscle cells and lens epithelial cells, and can act as a macrophage chemoattractant (Changsirikulchai et al.; Lokker et al.; Ray et al.; Uutela et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    STMC78222.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    PDGF-DD
  • Purity:
    ≥95%
  • Species:
    Human

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.