Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Mouse Recombinant SDF-1 alpha (CXCL12)

25 µG
Catalog # STMC78121.1
Marketsource
Mouse Recombinant SDF-1 alpha (CXCL12)
Catalog # STMC78121.1
Marketsource
CAS Number:  
Mouse Recombinant SDF-1 alpha (CXCL12)
Mouse Recombinant SDF-1 alpha (CXCL12)

25 µG

CAS Number:  
Price
kr2,199.00
Log in for availability
1 * 1 Each
Product Details & Documents

Stromal cell-derived factor 1 alpha (SDF-1α) is a member of the CXC group of chemokines that binds to the G-protein coupled receptor, CXCR4, to regulate migration, proliferation, differentiation, and survival of many cell types including hematopoietic stem cells, B cells, and T cells. It is produced by bone marrow stromal cells, osteoblasts, endothelial cells, and neuronal cells. SDF-1α was first identified as the pre-B-cell growth-stimulating factor (PBSF) in the mouse bone marrow-derived stromal cell line, PA6, in the growth of B cell precursors (Hayashi et al.). SDF-1α primarily regulates cell motility during development and adulthood, including the homing of hematopoietic stem cells and neutrophils to fetal bone marrow during ontogeny (Ara et al. 2003a) and the recruitment of endothelial progenitor cells from bone marrow during angiogenesis in adulthood (Zheng et al.). In addition to its role in hematopoiesis, the SDF-1α/CXCR4 signaling pathway is also essential for the homing of primordial germ cells to gonads (Ara et al. 2003b), the migration of granule cells in the cerebellum during neurogenesis (Zou et al.), and the migration of breast cancer cells to sites of metastasis (Muller et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    25 µG
  • Cat. No.:
    STMC78121.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    SDF-1 alpha (CXCL12)
  • Purity:
    ≥95%
  • Species:
    Mouse

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.