Cepharanthine 95%
Glass bottle, 250 mg
Catalog # 468912500.
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (15S,31R)-36,54-Dimethoxy-16,32-dimethyl-15,16,17,18,31,32,33,34-octahydro-2,6-dioxa-1(4,5)-[1,3]dioxolo[4,5-g]isoquinolina-3(7,1)-isoquinolina-5(1,3),7(1,4)-dibenzenacyclooctaphane, 6',12'-Dimethoxy-2,2'-dimethyl-6,7-(methylenebis(oxy))oxyacanthan, [14aS-(14aR*,26aS*)]-2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl- H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2′,3′:17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline, CEP, O-Methylcepharanoline, (14S,27R)-22,33-dimethoxy-13,28-dimethyl-2,5,7,20-tetraoxa-13,28-diazaoctacyclo[25.6.2.216,19.13,10.121,25.04,8.031,35.014,39]nonatriaconta-1(33),3(39),4(8),9,16(38),17,19(37),21,23,25(36),31,34-dodecaene
- CAS Number:481-49-2
- Formula:C₃₇H₃₈N₂O₆
- MDL Number:MFCD00210482
- Molecular Weight:606.7 g/mol
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:white to light yellow to beige to light orange crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Loss on drying:≤1.5 %
Specifications
- Pack type:Glass bottle
- Pk:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



