Cepharanthine 95%
Glass bottle, 1 g
Catalog # 468910010.
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (15S,31R)-36,54-Dimethoxy-16,32-dimethyl-15,16,17,18,31,32,33,34-octahydro-2,6-dioxa-1(4,5)-[1,3]dioxolo[4,5-g]isoquinolina-3(7,1)-isoquinolina-5(1,3),7(1,4)-dibenzenacyclooctaphane, 6',12'-Dimethoxy-2,2'-dimethyl-6,7-(methylenebis(oxy))oxyacanthan, [14aS-(14aR*,26aS*)]-2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl- H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2′,3′:17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline, CEP, O-Methylcepharanoline, (14S,27R)-22,33-dimethoxy-13,28-dimethyl-2,5,7,20-tetraoxa-13,28-diazaoctacyclo[25.6.2.216,19.13,10.121,25.04,8.031,35.014,39]nonatriaconta-1(33),3(39),4(8),9,16(38),17,19(37),21,23,25(36),31,34-dodecaene
- CAS Number:481-49-2
- Formula:C₃₇H₃₈N₂O₆
- MDL Number:MFCD00210482
- Molecular Weight:606.7 g/mol
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:white to light yellow to beige to light orange crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Loss on drying:≤1.5 %
Specifications
- Pack type:Glass bottle
- Pk:1 g



