DIGX RABBIT ANTI-DIGOXIGENIN LINKER (READY-TO-USE) 1 * 6 mL
Catalog # ENZOENZABS3036000
Supplier: ENZO LIFE SCIENCES
New Product
DIGX RABBIT ANTI-DIGOXIGENIN LINKER (READY-TO-USE) 1 * 6 mL
Catalog # ENZOENZABS3036000
Supplier: ENZO LIFE SCIENCES
New Product
Frequently Bought Together
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier: VWR Collection
Measuring scoops
Supplier: Brand
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Cell culture vessels, Corning® HYPERflask™ M, CellBIND® treated
Supplier: Corning
CellAdhereTrademark Fibronectin-Coated, 6-Well Flat-Bottom Plate 1 * 50 items
Supplier: STEMCELL Technologies
Reagent/centrifuge tubes, conical bottom, with push cap
Supplier: SARSTEDT
Screw cap microcentrifuge tubes, SuperClear®
Supplier: LABCON
BOTTLE PET 350ML RECT STERILE 0.5ML/18MG THIOSULPHATE RED DOME CAP 1 * 174 items
Supplier: LAB3 OWN PRODUCTION
BGER PP M/TRYKLG 500ML 1 * 1 Items
Supplier: NOLATO HERTILA
Micro-cavity plates, Corning® Elplasia®
Supplier: Corning
Microcentrifuge tubes, 0,6/1,5/1,7/2,0/5,0 ml, SnapLock, Axygen®
Supplier: Corning