Skip to Header
Skip to Main Content
Skip to Footer
Udvidet søgning
Udvidet søgning
SDS
Certifikater
Producentdata
0
0,00 kr
Order Entry
Log ind
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KECK MOUNTING SET KOMPLEET WITH SHORT SCREW 1 * 1 items
Catalog # SCOT286342802
Supplier:
DWK Life Sciences
Udvid billedet
KECK MOUNTING SET KOMPLEET WITH SHORT SCREW 1 * 1 items
Catalog # SCOT286342802
Supplier:
DWK Life Sciences
Frequently Bought Together
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR® Flasks, Round Bottom, with Standard Ground joint
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection