,,16 components: 100mg/l each of Al As Ba Ca Cd Cr Cu Fe Mg Mo Ni Pb Sb Se Sn Zn in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM106196L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together