,,13 components: 100mg/l each of Al Ba Ca Fe K Mg Mn Na P S Si Sr Ti in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM079341L5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together