Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

ACE-2

Plastic bottle, 1 mg
Catalog # AATB13555
Supplier:   AAT Bioquest
ACE-2
Catalog # AATB13555
Supplier:  AAT Bioquest
CAS Number:  
ACE-2
ACE-2

Plastic bottle, 1 mg

CAS Number:  
Price
2.240,00 kr
Log in for availability
1 * 1 mg
Product Details & Documents

ACE2 (angiotensin-converting enzyme 2) is a metalloproteinase that requires a divalent cation positioned at the active site in order to perform catalysis.

  • High flexibility
  • Higher throughput
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Cat. No.:
    AATB13555
  • Molecular formula:
    C₃₂H₃₆N₆O₁₂
  • Protein/peptide name:
    ACE-2
  • Grade:
    Molecular Biology Grade
  • Tested applications:
    Fluorescence
  • Molecular weight:
    696,7 g/mol

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.