Order Entry
Denmark
ContactUsLinkComponent
5x HF.E.35326 socket strip UK version 5x HF.I.35305 perforated shelf, powder-coated sheet steel, RAL 7035 light grey, load capacity=75 kg 1x EP.ML.34775 bottom collecting sump standard, powder-coated sheet steel, RAL 7035 light grey, capacity=2.9 lit 1 * 1 items
Catalog # MSGA41109
Supplier:  asecos
5x HF.E.35326 socket strip UK version 5x HF.I.35305 perforated shelf, powder-coated sheet steel, RAL 7035 light grey, load capacity=75 kg 1x EP.ML.34775 bottom collecting sump standard, powder-coated sheet steel, RAL 7035 light grey, capacity=2.9 lit 1 * 1 items
Catalog # MSGA41109
Supplier:  asecos
Frequently Bought Together