Order Entry
Denmark
ContactUsLinkComponent
Calcium Carbonate - Certified Reference Material for standardization of volumetric solutions CRM, ISO 17034 and ISO 17025 Conservation 60 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 50 g
Catalog # CMPLVS1271.50G
Calcium Carbonate - Certified Reference Material for standardization of volumetric solutions CRM, ISO 17034 and ISO 17025 Conservation 60 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 50 g
Catalog # CMPLVS1271.50G
Frequently Bought Together