Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium hydroxide 3% in aqueous solution

Sodium hydroxide 3% in aqueous solution

Glass bottle, 3 L
Catalog # AMPQ14720.1000-003
Corrosive
Supplier:   Ampliqon A/S
Sodium hydroxide 3% in aqueous solution
Catalog # AMPQ14720.1000-003
Supplier:  Ampliqon A/S
CAS Number:  1310-73-2
Sodium hydroxide 3% in aqueous solution
Sodium hydroxide 3% in aqueous solution

Glass bottle, 3 L

CAS Number:  1310-73-2
Price
4.310,00 kr
Log in for availability
1 * 3 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    3 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.