Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium hydroxide 3% in aqueous solution

Sodium hydroxide 3% in aqueous solution

Glass bottle, 1 L
Catalog # AMPQ14720.1000-001
Corrosive
Supplier:   Ampliqon A/S
Sodium hydroxide 3% in aqueous solution
Catalog # AMPQ14720.1000-001
Supplier:  Ampliqon A/S
CAS Number:  1310-73-2
Sodium hydroxide 3% in aqueous solution
Sodium hydroxide 3% in aqueous solution

Glass bottle, 1 L

CAS Number:  1310-73-2
Price
3.510,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications