23 components: 100mg/l each of Al Sb Ba B Cd Ca Cr Co Cu Fe Pb Li Mg Mn Mo Ni P K Si Na Ti V Zn in HNO3 5% HF 0.1% 1 * 100 mL
Catalog # CMPLRM014823L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together