Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TOC STANDARD SOLUTION 10000 MG/L 1 * 2 L
Catalog # CMPLTOC10000.L5
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
TOC STANDARD SOLUTION 10000 MG/L 1 * 2 L
Catalog # CMPLTOC10000.L5
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
7 components: 1000mg/l each of As Ba Be Se Te U V in HNO3 10% ,, CRM, ISO 17034 and ISO 17025,, Conservation 24 months. (12 months after opening),, Ambient Temp.,, 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Sol.,Per Acid 0.5%,500ML 1 * 1 items
Supplier: Epredia