ISOTHIAZOLINONE REAGENT DK5 1 * 65 mL
Catalog # LOVI56L046565
Supplier: Lovibond
Frequently Bought Together
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Visual colour kits, iron, MQuant®, Supelco®
Supplier: Merck
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Lactophenol blue solution stain (fungal), Sigma-Aldrich®
Supplier: Merck
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Sigma-Aldrich® Bouin's solution
Supplier: SIGMA ALDRICH MICROSCOPY
REA Control Antibody, human IgG1, APC, REAfinity 1 * 30 µG
Supplier: MILTENYI BIOTEC
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY