Order Entry
Denmark
ContactUsLinkComponent
4 components: Ca 200mg/l Cl- 200mg/l P 25mg/l S 100mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM101327L1
4 components: Ca 200mg/l Cl- 200mg/l P 25mg/l S 100mg/l in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 100 mL
Catalog # CMPLRM101327L1
Frequently Bought Together