[EN]SODIUM PALMITATE (U- C. 98%) 1 * 1 g
Catalog # CAMBCLM-6059-1
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection