Order Entry
Denmark
Orders LinkContactUsLinkComponent
Inserts

Inserts

Insert for the optimum loading of up to 8 trays or cassettes, equipped with 9 holders, spacing: 40 mm, PWD 8531, PWD 8532, 103×389×110 mm
Catalog # MIEL11017170
Supplier:   MIELE
Inserts
Catalog # MIEL11017170
Supplier:  MIELE
Inserts
Inserts

Insert for the optimum loading of up to 8 trays or cassettes, equipped with 9 holders, spacing: 40 mm, PWD 8531, PWD 8532, 103×389×110 mm

Price
580,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • B×D×H udv.:
    103×389×110 mm
  • Beskrivelse:
    Insert for the optimum loading of up to 8 trays or cassettes, equipped with 9 holders, spacing: 40 mm
  • Seq:
    58
  • Til:
    PWD 8531, PWD 8532
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.