Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Log ind
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1 component: Ethyl-tert-butylether (ETBE) [CAS:637-92-3] 2000mg/l in Met 1 * 5 Ampoul
Catalog # CMPLRD0106551.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
1 component: Ethyl-tert-butylether (ETBE) [CAS:637-92-3] 2000mg/l in Met 1 * 5 Ampoul
Catalog # CMPLRD0106551.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach