Furnace ashing lv15/11/c550 tMax1100 with FlaP Door anD therMocouPle hole in the Furnace Door 1 * 1 items
Catalog # NABELV155K11NOZ1
Supplier: NABERTHERM
Furnace ashing lv15/11/c550 tMax1100 with FlaP Door anD therMocouPle hole in the Furnace Door 1 * 1 items
Catalog # NABELV155K11NOZ1
Supplier: NABERTHERM
Frequently Bought Together
Overhead stirrer tools, dissolver stirrers
Supplier: IKA
Accessories for Retsch sieves
Supplier: Retsch GmbH
Magnetic stirrer, Topolino
Supplier: IKA
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
High volume magnetic stirrers
Supplier: VELP SCIENTIFIC
VWR®, Disposable Pestles, Microtubes and Cordless Motor for Pellet Mixing
Supplier: VWR Collection
Magnetic stirrers, MICROSTIRRER
Supplier: VELP SCIENTIFIC
Dual action shaker, KL 2
Supplier: Edmund Bühler GmbH
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Accessories for tube mill control
Supplier: IKA
Vortex mixer, VORTEX 2
Supplier: IKA
VWR®, Stainless Steel Test Tube Racks
Supplier: VWR Collection
VWR® VOS 40 digital & VOS 60 control, Electronic Overhead Stirrers
Supplier: VWR Collection
Blocks for IKA digital dry block heaters
Supplier: IKA
Overhead stirrer tools, centrifugal stirrers
Supplier: IKA
Thermoshakers for microtubes and PCR plates, PHMT series
Supplier: GRANT INSTRUMENTS