Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Flask, volumetric, capacity 250ML, NS 14/23, class A, conformity certified 1 * 2 items
Catalog # HIRS2820286
Supplier:
HIRSCHMANN
Expand image
Flask, volumetric, capacity 250ML, NS 14/23, class A, conformity certified 1 * 2 items
Catalog # HIRS2820286
Supplier:
HIRSCHMANN
Frequently Bought Together
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Volumetric flasks, USP, class A, blue markings, BLAUBRAND®
Supplier: Brand
Sand, TECHNICAL, industrial, washed
Supplier: VWR Chemicals
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Melting point determination tubes
Supplier: HIRSCHMANN