Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CF24
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CF24
Supplier: Sartorius Balances
Frequently Bought Together
ATTACHMENT STANDARD VG 3.1 1 * 1 items
Supplier: IKA
VWR®, Vortex Mixers, Heavy Duty, Analogue and Digital
Supplier: VWR Collection
Stirrers, propeller, with four paddles
Supplier: BOCHEM
Safety Locking Devices
Supplier: Retsch GmbH
Accessories for shakers
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for shakers, KS 260 and HS 260
Supplier: IKA
Blocks for IKA digital dry block heaters
Supplier: IKA
Overhead stirrer multi mixer up to 20 1 * 1 items
Supplier: GRANT INSTRUMENTS