Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
SARBMCA36S3S002
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
SARBMCA36S3S002
Frequently Bought Together
VWR®, Mini vortex mixer
: VWR Collection
Vortex mixers, MS 3
: IKA
Vortex mixers, Vortex-Genie®
: SCIENTIFIC INDUSTRIES ELMIS
Accessories for MB series
: OHAUS
Gas cartridges, Campingaz®
: CAMPING GAZ
Stirrer bearings, PTFE
: Bohlender
VWR®, Vortex Mixer
: VWR Collection
VWR® DRY-Line® Prime, Drying Ovens with Natural Convection
: VWR Collection
VWR® VWB2, Water Baths
: VWR Collection
VWR® lab dancer, Vortex Mixer
: VWR Collection
Cartridge 220G for cartridge burner
: USBECK Laborgeräte
Accessories for MA series
: Sartorius Balances
Accessories for Retsch sieves
: Retsch GmbH
Bunsen burner, cartridge type
: USBECK Laborgeräte